Learn More
Grainger Fork End 304 SS Cable Size 5/32
Catalog No. 502767977
Encompass_Preferred
The Loos & Co. 1/16" Wire Rope Clevis Fitting is made from drop forged steel for durability. It has a working load limit of 110 lbs and is rated for general purpose use. The clevis fitting accepts 1/16" wire rope and measures 1-1/4" x 5/8" x 1/2" overall. It features a clevis on one end and a threaded eye on the other end to accept a wire rope. This wire rope clevis fitting is ideal for creating an adjustable wire rope assembly for lifting, pulling, and securing applications. It is zinc plated for corrosion resistance.
Catalog No. 50-276-7977 Supplier Grainger Supplier No. 16X652
The Loos & Co. 1/16" Wire Rope Clevis Fitting is made from drop forged steel for durability. It has a working load limit of 110 lbs and is rated for general purpose use. The clevis fitting accepts 1/16" wire rope and measures 1-1/4" x 5/8" x 1/2" overall. It features a clevis on one end and a threaded eye on the other end to accept a wire rope. This wire rope clevis fitting is ideal for creating an adjustable wire rope assembly for lifting, pulling, and securing applications. It is zinc plated for corrosion resistance.