Learn More
Grainger Loos and Co Inc. Fork End Fitting, Stainless Steel, 7/32 in. Cable Size, 1 ea
Catalog No. 502767054
Encompass_Preferred
This fork end fitting is manufactured from 304 grade stainless steel in accordance with MIL-DTL-781 standards for critical wire rope applications. Designed for use with stainless steel and galvanized cable, this swage-type fitting is suitable for lifting operations.
  • Constructed from durable stainless steel to meet MIL-DTL-781 specifications
  • Compatible with 7/32 in. wire rope diameter
  • Features a 5,600 lb. minimum breaking strength
  • Designed for swage installation with 5/16 in. pin diameter compatibility
  • DFARS compliant and manufactured in the USA
Catalog No. 50-276-7054 Supplier Grainger Supplier No. 16X654
This fork end fitting is manufactured from 304 grade stainless steel in accordance with MIL-DTL-781 standards for critical wire rope applications. Designed for use with stainless steel and galvanized cable, this swage-type fitting is suitable for lifting operations.
  • Constructed from durable stainless steel to meet MIL-DTL-781 specifications
  • Compatible with 7/32 in. wire rope diameter
  • Features a 5,600 lb. minimum breaking strength
  • Designed for swage installation with 5/16 in. pin diameter compatibility
  • DFARS compliant and manufactured in the USA