Learn More
Grainger Fork End 304 SS Cable Size 9/32
Catalog No. 502768009
Encompass_Preferred
The Loos Wire Rope Clevis Fitting is a forged steel clevis with a 3/4" diameter hole that can accommodate wire rope up to 5/8" diameter. It is rated up to 9 tons working load limit. The clevis has a black oxide finish and includes a cotter pin for securing the wire rope. This heavy duty clevis fitting is ideal for creating an attachment point for wire rope in rigging applications. Made in the USA by Loos & Co., this clevis provides a safe and durable termination for wire rope.
Catalog No. 50-276-8009 Supplier Grainger Supplier No. 16X656
The Loos Wire Rope Clevis Fitting is a forged steel clevis with a 3/4" diameter hole that can accommodate wire rope up to 5/8" diameter. It is rated up to 9 tons working load limit. The clevis has a black oxide finish and includes a cotter pin for securing the wire rope. This heavy duty clevis fitting is ideal for creating an attachment point for wire rope in rigging applications. Made in the USA by Loos & Co., this clevis provides a safe and durable termination for wire rope.