Learn More
Grainger Fork End 316 SS Cable Size 1/8
Catalog No. 502767140
Encompass_Preferred
The Loos Wire Rope Clevis Fitting is a 1/16" clevis fitting made of forged steel with a zinc finish. It has a working load limit of 110 lbs and is rated for general purpose use. The clevis fitting accepts 1/16" diameter wire rope and features a threaded pin with cotter pin for securing the wire rope. Overall length is 1-1/4" with a 5/8" hole diameter. This clevis fitting is suitable for light duty applications like securing guy wires, attaching pulleys and creating adjustable wire rope lengths. Made in the USA by Loos & Co.
Catalog No. 50-276-7140 Supplier Grainger Supplier No. 16X660
The Loos Wire Rope Clevis Fitting is a 1/16" clevis fitting made of forged steel with a zinc finish. It has a working load limit of 110 lbs and is rated for general purpose use. The clevis fitting accepts 1/16" diameter wire rope and features a threaded pin with cotter pin for securing the wire rope. Overall length is 1-1/4" with a 5/8" hole diameter. This clevis fitting is suitable for light duty applications like securing guy wires, attaching pulleys and creating adjustable wire rope lengths. Made in the USA by Loos & Co.