Learn More
Grainger Fork End 316 SS Cable Size 3/16
Catalog No. 502765653
Encompass_Preferred
The Grainger 316 stainless steel fork end fittings for wire rope are designed to terminate wire rope for lifting applications. They are made from 316 stainless steel for corrosion resistance and feature a threaded stud with nut for attaching to equipment. Fork openings range from 5/16" to 1-1/4" to accommodate different wire rope diameters. Load ratings vary by size from 1,250 lbs to 9,800 lbs. Fork end fittings provide a secure method to create an eye on wire rope for attachment to hooks, shackles, and other hardware.
Catalog No. 50-276-5653 Supplier Grainger Supplier No. 16X662
The Grainger 316 stainless steel fork end fittings for wire rope are designed to terminate wire rope for lifting applications. They are made from 316 stainless steel for corrosion resistance and feature a threaded stud with nut for attaching to equipment. Fork openings range from 5/16" to 1-1/4" to accommodate different wire rope diameters. Load ratings vary by size from 1,250 lbs to 9,800 lbs. Fork end fittings provide a secure method to create an eye on wire rope for attachment to hooks, shackles, and other hardware.