Learn More
Grainger Fork End 316 SS Cable Size 3/32
Catalog No. 502767138
Encompass_Preferred
The Loos Wire Rope Clevis Fitting is a 1/16" clevis fitting made of forged steel with a zinc finish. It has a working load limit of 250 lbs and is rated for general purpose use. The clevis has a 1/16" hole size and is compatible with 1x19 and 7x7 wire rope. Overall length is 1-1/4" and weight is 0.02 lbs. This clevis fitting is often used for light duty suspension, hanging or securing applications. It should be inspected periodically for wear and damage.
Catalog No. 50-276-7138 Supplier Grainger Supplier No. 16X659
The Loos Wire Rope Clevis Fitting is a 1/16" clevis fitting made of forged steel with a zinc finish. It has a working load limit of 250 lbs and is rated for general purpose use. The clevis has a 1/16" hole size and is compatible with 1x19 and 7x7 wire rope. Overall length is 1-1/4" and weight is 0.02 lbs. This clevis fitting is often used for light duty suspension, hanging or securing applications. It should be inspected periodically for wear and damage.