Learn More
Grainger Fork End 316 SS Cable Size 7/32
Catalog No. 502767903
Encompass_Preferred
The Loos & Co. Wire Rope Clevis Fitting is a 1/16" clevis made of drop forged carbon steel with a zinc finish. It has a working load limit of 110 lbs and is rated for general purpose use. The clevis pin is held in place by a cotter pin. This small clevis fitting is suitable for light duty applications requiring a secure method of attaching wire rope to various connection points. With its compact size, it can be used in tight spaces. The zinc finish provides corrosion resistance. Overall, this is a versatile, general purpose wire rope clevis fitting for light duty applications.
Catalog No. 50-276-7903 Supplier Grainger Supplier No. 16X663
The Loos & Co. Wire Rope Clevis Fitting is a 1/16" clevis made of drop forged carbon steel with a zinc finish. It has a working load limit of 110 lbs and is rated for general purpose use. The clevis pin is held in place by a cotter pin. This small clevis fitting is suitable for light duty applications requiring a secure method of attaching wire rope to various connection points. With its compact size, it can be used in tight spaces. The zinc finish provides corrosion resistance. Overall, this is a versatile, general purpose wire rope clevis fitting for light duty applications.