Learn More
Grainger 3M Insulated Locking Fork Terminal, Vinyl, 1/4 in. Stud, 12-10 AWG, 500/Case
Catalog No. 502793820
Encompass_Preferred
These locking fork terminals are designed for reliable and secure connections in electrical systems. Featuring a vinyl-insulated, brazed seam barrel, these terminals accommodate wire gauges ranging from 12 AWG to 10 AWG. The locking fork configuration ensures the terminal remains securely on the stud, while the durable construction is rated for up to 600 V.
  • Brazed seam barrel construction for superior performance
  • Locking fork configuration for improved connection security
  • Vinyl insulation for enhanced safety and durability
  • Tin contact finish for reliable conductivity
  • Compatible with 1/4 in. stud sizes
  • Designed for 12-10 AWG wire range
Catalog No. 50-279-3820 Supplier Grainger Supplier No. 1A089
These locking fork terminals are designed for reliable and secure connections in electrical systems. Featuring a vinyl-insulated, brazed seam barrel, these terminals accommodate wire gauges ranging from 12 AWG to 10 AWG. The locking fork configuration ensures the terminal remains securely on the stud, while the durable construction is rated for up to 600 V.
  • Brazed seam barrel construction for superior performance
  • Locking fork configuration for improved connection security
  • Vinyl insulation for enhanced safety and durability
  • Tin contact finish for reliable conductivity
  • Compatible with 1/4 in. stud sizes
  • Designed for 12-10 AWG wire range