Learn More
Grainger Approved Vendor Tail Light, Incandescent, 12V DC, Metal And Plastic, 1 ea
Catalog No. 502779780
Encompass_Preferred
This combination tail light provides reliable stop, turn, and tail signaling for forklift trucks. Designed for industrial use, this rectangular unit features a durable metal housing and a plastic lens, ensuring visibility and performance in demanding environments.
  • Functions as a brake, reverse, and running light
  • Operating voltage of 12V DC with 1.8 A current draw
  • Meets SAE J845 standards
  • Includes wire leads for electrical connection
  • Surface mount installation with screws
  • IP65 rated for protection against dust and water
  • Dimensions: 25.4 cm (10 in.) width by 10.2 cm (4 in.) height by 7.6 cm (3 in.) depth
Catalog No. 50-277-9780 Supplier Grainger Supplier No. 19ND87
This combination tail light provides reliable stop, turn, and tail signaling for forklift trucks. Designed for industrial use, this rectangular unit features a durable metal housing and a plastic lens, ensuring visibility and performance in demanding environments.
  • Functions as a brake, reverse, and running light
  • Operating voltage of 12V DC with 1.8 A current draw
  • Meets SAE J845 standards
  • Includes wire leads for electrical connection
  • Surface mount installation with screws
  • IP65 rated for protection against dust and water
  • Dimensions: 25.4 cm (10 in.) width by 10.2 cm (4 in.) height by 7.6 cm (3 in.) depth