Learn More
Grainger Grainger Forklift Forks, 36 in. Length, 5000 lb. Capacity, Pair
Catalog No. 502782385
Encompass_Preferred
These forklift forks are designed for material handling applications, providing a robust solution for lifting and moving heavy loads. Manufactured to meet ITA Class 2 standards, these forks ensure compatibility and reliability for industrial equipment.
  • Load capacity of 5,000 lb.
  • Fork length of 36 in.
  • Fork width of 4 in.
  • Thickness of 1 3/4 in.
  • Constructed for high durability and performance
  • Compliant with ITA Class 2 standards
Catalog No. 50-278-2385 Supplier Grainger Supplier No. 19ND76
These forklift forks are designed for material handling applications, providing a robust solution for lifting and moving heavy loads. Manufactured to meet ITA Class 2 standards, these forks ensure compatibility and reliability for industrial equipment.
  • Load capacity of 5,000 lb.
  • Fork length of 36 in.
  • Fork width of 4 in.
  • Thickness of 1 3/4 in.
  • Constructed for high durability and performance
  • Compliant with ITA Class 2 standards