Learn More
Grainger Approved Vendor Fork, 4 in. Width, 48 in. Length, 5,000 lb. Capacity, 1 Pair
Catalog No. 502779784
Encompass_Preferred
These forklift forks are designed to attach to a carriage for lifting pallets and various types of cargo. They provide essential support for specialized applications, including drum handling, sweeping, and lifting items like carpet rolls. Constructed for durability and heavy-duty performance, these forks are built to meet demanding operational needs in industrial settings.
  • Maximum load capacity of 5,000 lb
  • Dimensions include 4 in. width, 48 in. length, and 1 3/4 in. thickness
  • Compliant with ITA Class 2 standards
  • Supplied as a pair
Catalog No. 50-277-9784 Supplier Grainger Supplier No. 19ND78
These forklift forks are designed to attach to a carriage for lifting pallets and various types of cargo. They provide essential support for specialized applications, including drum handling, sweeping, and lifting items like carpet rolls. Constructed for durability and heavy-duty performance, these forks are built to meet demanding operational needs in industrial settings.
  • Maximum load capacity of 5,000 lb
  • Dimensions include 4 in. width, 48 in. length, and 1 3/4 in. thickness
  • Compliant with ITA Class 2 standards
  • Supplied as a pair