Learn More
Formlabs Inc Form 3 Resin Tank
SDP
Catalog No. NC1951633
Encompass
The Formlabs Form 3 Resin Tank is a replacement resin tank for the Form 3 and Form 3L 3D printers. It is made from injection molded polycarbonate for durability and chemical resistance. The tank has a 250 mL resin capacity and an integrated RFID chip that allows the printer to automatically detect and configure settings for this tank. The tank's proprietary low force peel mechanism enables effortless resin separation after prints. Overall, the Form 3 Resin Tank is an official replacement part made specifically for the Form 3 and Form 3L 3D printers.
Catalog No. NC1951633 Supplier Formlabs Inc Supplier No. RTF30201
The Formlabs Form 3 Resin Tank is a replacement resin tank for the Form 3 and Form 3L 3D printers. It is made from injection molded polycarbonate for durability and chemical resistance. The tank has a 250 mL resin capacity and an integrated RFID chip that allows the printer to automatically detect and configure settings for this tank. The tank's proprietary low force peel mechanism enables effortless resin separation after prints. Overall, the Form 3 Resin Tank is an official replacement part made specifically for the Form 3 and Form 3L 3D printers.