Learn More
Formlabs Inc FORM 3L RESIN TANK V3
SDP
Catalog No. NC2590365
Encompass
Low Force Stereolithography (LFS)™ 3D printing uses linear illumination and a flexible film tank to turn liquid resin into flawless parts. A key component of the LFS process, the strong dual-layer film at the bottom of the Form 3L Resin Tank allows for a dynamic and precise print process, scaled up for large format printing.For use with Form 3L and Form 3BL.Notice: Form 3L Resin Tank V3 offers an improved print success rate in some cases compared to the previous version of the Form 3L Resin Tank V2. Notice: Elastic 50A Resin is not yet validated with Form 3L Resin Tank V3. We expect to complete validation of Elastic 50A Resin and Form 3L Resin Tank V3 by the end of Q4 2023.
Catalog No. NC2590365 Supplier Formlabs Inc Supplier No. RTF3L03
Low Force Stereolithography (LFS)™ 3D printing uses linear illumination and a flexible film tank to turn liquid resin into flawless parts. A key component of the LFS process, the strong dual-layer film at the bottom of the Form 3L Resin Tank allows for a dynamic and precise print process, scaled up for large format printing.For use with Form 3L and Form 3BL.Notice: Form 3L Resin Tank V3 offers an improved print success rate in some cases compared to the previous version of the Form 3L Resin Tank V2. Notice: Elastic 50A Resin is not yet validated with Form 3L Resin Tank V3. We expect to complete validation of Elastic 50A Resin and Form 3L Resin Tank V3 by the end of Q4 2023.