Learn More
Formlabs Inc FORM CURE
SDP
Supplier:  Formlabs Inc FHCU01
Encompass
Post-curing maximizes material properties for stereolithography prints, improving parts’ strength and performance. Form Cure precisely combines temperature and 405 nm light to consistently post-cure Form 2 prints.
Catalog No. NC1650536