Learn More
Electron Microscopy Sciences Cold Embedding Form Mold, 40 mm
SDP
Catalog No. 501929681
Encompass_Preferred
The form mold is a two part mounting cup, designed to be as user-friendly as possible. It has practical handles for removing of the lid and the cylinder is slightly conical for easy removal of the mounts.
Diameter: 40 mm
Catalog No. 50-192-9681 Supplier Electron Microscopy Sciences Supplier No. 5101203
The form mold is a two part mounting cup, designed to be as user-friendly as possible. It has practical handles for removing of the lid and the cylinder is slightly conical for easy removal of the mounts.
Diameter: 40 mm