Learn More
Description
Obtain the convenience, reassurance and reliability required in the event of a power or mechanical failure with the Thermo Scientific™ Forma™ CO2 Backup Systems for ultra low temperature freezers, a must for critical storage applications.
- Temperature is adjustable in 1°C increments to -75°C
- Digital display indicates actual cabinet temperature
- Indicator lights illuminate when system is activated
- Liquid flow shuts off automatically when freezer door or lid is opened
- System powered by rechargeable battery
- Low-battery light indicates when battery requires charging
- Additional indicator lights when cylinder is empty
Ordering Information:
Some models require freestanding system; others accept built-in systems. Specify freezer model and voltage when ordering.
Includes:
All necessary hardware except CO2 cylinder (must be ordered separately)
Specifications
Specifications
| Description | High-pressure Hose |
| Type | Accessory |
| Temperature Range | -75°C |
| Style | Built-in |
| Defrost | Automatic |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.