Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Forma™ Freezer Replacement Shelves

Catalog No. 13994054 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
17.3 cu. ft. Freezers
Type:
Shelf Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
13-994-054 17.3 cu. ft. Freezers Shelf Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-994-054 Supplier Thermo Scientific™ Supplier No. 224135
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.

  • Stainless steel
  • Kits include mounting hardware
TRUSTED_SUSTAINABILITY

Specifications

Type Shelf Kit
Capacity (English) 17.3 cu. ft.
Capacity (Metric) 489.8 L
Style Upright
Color Silver
For Use With (Equipment) 17.3 cu. ft. Freezers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.