Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Forma™ Side-Access Box Racks

Catalog No. 920009 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
Boxes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
920009 Boxes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 920009 Supplier Thermo Scientific™ Supplier No. 920009

missing translation for 'validMainframeMsg'

Access ultra-low storage in upright freezers when sliding drawer racks are not preferred with these side-access box racks, available with and without boxes.

Access ultra-low storage in upright freezers when sliding drawer racks are not preferred with Thermo Scientific™ Forma™ Side-Access Box Racks, available with and without boxes.

  • Use racks in single-door, upright freezers.
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Color Silver
Holds Boxes
Material Stainless Steel
Positions 9
Shape Rectangular
Stackable Yes
Sterility Non-sterile
For Use With (Equipment) 13 cu. ft. (368L) capacity upright freezers
Product Type Freezer Storage Rack
Height (Metric) 55.8 cm
Width (Metric) 14 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.