Learn More
Andwin Scientific FORMAGO FORMALDEHYDE ANALY 1KT
SDP
Catalog No. 501125960
Encompass_Preferred
The Tissue-Tek FormaGO formaldehyde analysis kit allows consistent results and leverages a two-year shelf life, which improves inventory management. It includes two 25 mL reagent bottles and 100 test strips. CAS # 1310-73-2.
Catalog No. 50-112-5960 Supplier Andwin Scientific Supplier No. 10852348
The Tissue-Tek FormaGO formaldehyde analysis kit allows consistent results and leverages a two-year shelf life, which improves inventory management. It includes two 25 mL reagent bottles and 100 test strips. CAS # 1310-73-2.