Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Formal-Fixx™

Catalog No. 6764255 Shop All Epredia Products
Change view
Click to view available options
Quantity:
3.8 L concentrate (19 L total)
Product Type:
Fixative
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
6764255 3.8 L concentrate (19 L total) Fixative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6764255 Supplier Epredia™ Supplier No. 6764255
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acquire amazing results with Epredia™ Formal-Fixx™, which is excellent for immunochemistry.

  • Enhances H&Es and special stains
  • Minimizes the need to enzyme digest
  • Reduces protein cross-linking
  • Produces well-defined nuclear detail

Specifications

Product Type Fixative
Quantity 3.8 L concentrate (19 L total)
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.