Learn More
Electron Microscopy Sciences Formalde-Fresh Low Odor Solution, Histology Grade 4 LT
SDP
Catalog No. 50980481
Encompass_Preferred
A specially formulated odor-mask which neutralizes the formalin odor without affecting subsequent tissue processing, handling, or staining. Low odor, ready-to-use 10% formalin solution. May be used as a fixative or preservative of specimens. Its shelf life is 24 months.
Grade: Histology Grade
Catalog No. 50-980-481 Supplier Electron Microscopy Sciences Supplier No. 1567804
A specially formulated odor-mask which neutralizes the formalin odor without affecting subsequent tissue processing, handling, or staining. Low odor, ready-to-use 10% formalin solution. May be used as a fixative or preservative of specimens. Its shelf life is 24 months.
Grade: Histology Grade