Learn More
Electron Microscopy Sciences Formalde-Fresh Low Odor Solution, Histology Grade 4 LT
SDP
Supplier:  Electron Microscopy Sciences 1567801
Encompass_Preferred
A specially formulated odor-mask which neutralizes the formalin odor without affecting subsequent tissue processing, handling, or staining. Low odor, ready-to-use 10% formalin solution. May be used as a fixative or preservative of specimens. Its shelf life is 24 months.
Grade: Histology Grade
Catalog No. 50-980-480