Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 10% (v/v) (non-buffered), Histological Fixative, Ricca Chemical
SDP

Catalog No. R3180000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Capacity:
500 mL
1 L
4 L
10 L
20 L
Packaging Type:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Packaging Type
R3180000500 500 mL Natural Poly Bottle
3180-32 1 L Natural Poly Bottle
3180-1 4 L Natural Poly Bottle
3180-2-5 10 L Cubitainer
3180-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R3180000500 Supplier Ricca Chemical Company Supplier No. 318016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Capacity 500 mL
Color Colorless
Packaging Type Natural Poly Bottle
For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Formaldehyde
Solution Type Formaldehyde
Form Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.