Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 10% (v/v), Buffered, pH 7.4, Carson-Millonig Formulation, Ricca Chemical
SDP

Catalog No. 31915 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3191-5 20 L Cubitainer
3191-32 1 L Natural Poly Bottle
3191-1 4 L Natural Poly Bottle
3191-2-5 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 3191-5 Supplier Ricca Chemical Company Supplier No. 31915
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tissue Fixative for Light and Electron Microscopy

Specifications

Physical Form Liquid
Chemical Name or Material Formaldehyde
Name Note pH 7.4, Carson-Millonig Formulation
CAS 7732-18-5, 50-00-0, 67-56-1, 7664-38-2, 1310-73-2
CAS Min % 86.0, 9.2, 1.8, 0.1
CAS Max % 89.0, 9.57, 2.0, 0.9
Packaging Cubitainer
Treatment(s) Buffered
Color Colorless
pH 7.4
Quantity 20 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 10% (v/v)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.