Learn More
Tousimis Researc Corporation Formaldehyde 20%, 100mL
SDP
Catalog No. NC9998644
Encompass
Formaldehyde 20%, 100mL, No preservatives. Ready-to-use in your research laboratory as a primary or complimentary fixative of biological tissue-samples for electron, light microscopy, or other laboratory applications.
Catalog No. NC9998644 Supplier Tousimis Researc Corporation Supplier No. 1008B
Formaldehyde 20%, 100mL, No preservatives. Ready-to-use in your research laboratory as a primary or complimentary fixative of biological tissue-samples for electron, light microscopy, or other laboratory applications.