Learn More
AdipoGen Formaldehyde-2 4-DNPH
SDP
Supplier:  AdipoGen CDXF0382M010
Encompass_Preferred
Chemical. CAS 1081-15-8. Formula C7H6N4O4. Molecular Weight 210.15. Formaldehyde-2,4-DNPH can be used as reference compound for the determination of the analyte in air samples, food and feed products and aquatic products by various chromatography techniques.
Catalog No. 50-256-0852