Learn More
Tousimis Researc Corporation Formaldehyde 20%, 10x10mL
SDP
Catalog No. NC9611804
Encompass
FORMALDEHYDE,20% aqueous,10x10ml ampules,ultrapure,No preservatives,Ready-to-use in your research laboratory as a primary or complimentary fixative of biological tissue-samples for electron,light microscopy,or other laboratory applications
Catalog No. NC9611804 Supplier Tousimis Researc Corporation Supplier No. 1008A
Add to Cart
Add to Cart
FORMALDEHYDE,20% aqueous,10x10ml ampules,ultrapure,No preservatives,Ready-to-use in your research laboratory as a primary or complimentary fixative of biological tissue-samples for electron,light microscopy,or other laboratory applications