Learn More
Bioworld Formaldehyde, 500 mL
Supplier:  Bioworld 217300022
Encompass_Preferred
- 37% solution - Biotechnology Grade Scientific Chemical Name: H2CO Acidity(%)- <=0.03 Color (APHA)- <=10 Formaldehyde(%)- 37.0-37.5 Methanol(%)- 10.5-13.0
Catalog No. 50-199-029