Learn More
Electron Microscopy Sciences Formaldehyde, 37%, Microfiltered (Formalin Microfiltered), 50-00-0, 10x10ml
SDP
Supplier:  Electron Microscopy Sciences 15686
Encompass_Preferred
Molecular Formula CH2O, Molecular Weight 30.026, Formaldehyde, 37% has been microfiltered with a 0.45 micron membrane to make it suitable as an EM fixative when a methanol free solution is not necessary.
Catalog No. 50-980-485
 Restricted Item