Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, 8% (v/v) (non-buffered), Histological Fixative, Ricca Chemical
SDP

Catalog No. 317832 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
31-783-2 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-783-2 Supplier Ricca Chemical Company Supplier No. 317832
 Restricted Item
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Formaldehyde
Name Note 8% (v/v) Aqueous Solution (non-buffered), Histological Fixative
CAS 7732-18-5, 50-00-0, 67-56-1
Color Clear colorless
Boiling Point Approximately 100°C
Packaging Natural Poly Bottle
Quantity 1 L
Percent Purity 8% v/v
Grade Laboratory
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.