Learn More
Cayman Chemical Formaldehyde Dehydrogenase P
SDP
Catalog No. 502890533
Encompass_Preferred
Source: Active recombinant N-terminal His-tagged protein expressed in E. coli • MW: 44.6 kDa
Catalog No. 50-289-0533 Supplier Cayman Chemical Supplier No. 10352100 UG
Source: Active recombinant N-terminal His-tagged protein expressed in E. coli • MW: 44.6 kDa