Learn More
Arbor Assays Formaldehyde Detection Kit (1 Plate)
SDP
Supplier:  Arbor Assays K001F1
Encompass_Preferred
The DetectX Formaldehyde (HCHO) Fluorescent Detection Kit is designed to quantitatively measure formaldehyde present in buffer, tissue culture media and urine samples.
Catalog No. 50-227-8375