Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Formaldehyde dimethyl acetal

Catalog No. 47649250MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-649-250MLF 250 mL
47-649-1LF 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47-649-250MLF Supplier Sigma Aldrich Supplier No. 47649250MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Formaldehyde dimethyl acetal (fda, fdma, fadma, dimethoxymethane, dmm, methylal) is a biodegradable dimethyl acetal. it has been synthesized by condensing formaldehyde with methanol in the presence of acid catalyst. it is ampiphilic in nature with low viscosity, surface tension and boiling point. it is a flammable, highly volatile solvent with excellent dissolving power. it is reported to form trimethylorthoformate by anodic methoxylation in basic methanol.formaldehyde dimethyl acetal may be used in the following studies:
  • Synthesis of methoxymethyl (mom) ethers
  • As an external cross-linker to form microporous polymers
  • Dehydration of biological samples for scanning electron microscopy (sem)

Specifications

CAS 109-87-5
Density 0.860 g/mL (at 20°C (literature))
Molecular Formula C3H8O2
Linear Formula CH2(OCH3)2
MDL Number MFCD00008495
Quantity 250 mL
Synonym Dimethoxymethane; Methylal
Molecular Weight (g/mol) 76.09
Percent Purity ≥99.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.