Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Formaldehyde Monitor

Catalog No. p-177010 Shop All 3M Products
Change view
Click to view available options
Product Type:
Formaldehyde Monitor
Formaldehyde Monitor only
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17-985-363C Formaldehyde Monitor only
10-030-84 Formaldehyde Monitor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-985-363C Supplier 3M™ Supplier No. 7000051779
This item is not returnable. View return policy

Complies with OSHA formaldehyde standard

Valuable for use in chemical, textile, and pulp and paper industries; histology; healthcare.

  • Sensitivity of 0.8ppm-hours (0.1ppm over an eight-hour period) Upper exposure limit of 40ppm-hours (5ppm over an eight-hour period)
  • Available as a monitor only or with prepaid analysis
  • Not for 15-minute STEL (short-term exposure limit) sampling. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product. Misuse may result in sickness or death.
  • 3M Catalog# 3720 , 3M Catalog# 3721

Compliance

Exceeds OSHA accuracy requirements for eight-hour sampling.

TRUSTED_SUSTAINABILITY

Specifications

Color Purple
For Use With (Application) Valuable for use in chemical, textile and pulp and Paper industries, histology, healthcare
Certifications/Compliance OSHA
Detectable Analytes Formaldehyde
Measurement Range 40 ppm.Hour
Product Type Formaldehyde Monitor only
Model 3721
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.