Learn More
Hach Company Formaldehyde 1 Reagent Powder Pillows, pk/100
Supplier:  Hach Company 2182999
Encompass
For high-range formaldehyde determination by drop-count titration. Replacement reagent for FM-1 test kit (2183100). Pack of 100 Powder Pillows.
Catalog No. 50-199-5515