Learn More
Sigma Aldrich Fine Chemicals Biosciences formaldehyde solutin cont500ML
Catalog No. 502900011
Encompass
This product is 37wt% SOLUTIONon of formaldehyde in water. It contains 10-15% methanol as stabilizer.
Catalog No. 50-290-0011 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F1635500ML
This product is 37wt% SOLUTIONon of formaldehyde in water. It contains 10-15% methanol as stabilizer.