Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde Solution, 10%, w/v in aqueous Phosphate Buffer, Macron Fine Chemicals™

Catalog No. MKH1214 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-H121-4 4 L Poly Bottle
MK-H121-1 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MK-H121-4 Supplier Avantor Supplier No. H12108

missing translation for 'validMainframeMsg'

Chemical Identifiers

Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

CAS 50-0-0
Packaging Poly Bottle
Quantity 4 L
Chemical Name or Material Formaldehyde Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.