Learn More
Supply Solutions Avantor Performance Materials - Formaldehyde Solution. Size: 20L; Grade: Ar (Acs); Concentration: 36.5-38%.; Size: 20L
Catalog No. 50878436
Encompass_Preferred
Formaldehyde Solution. Grade: Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs. Molar Mass: 30.03 g/mol. Manufacturer Name: EMD Millipore. Manufacturer Part No: FX0415-6; Size: 20L; Grade: AR (ACS); Concentration: 36.5-38%; CAS: 50-00-0; Hazardous: Y; Container Material: Cubitainer; Formula: HCHO; Formula Weight: 30.03; Suitable for Histological Applications
Catalog No. 50-878-436 Supplier Supply Solutions Supplier No. MACRON 501619
Formaldehyde Solution. Grade: Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs. Molar Mass: 30.03 g/mol. Manufacturer Name: EMD Millipore. Manufacturer Part No: FX0415-6; Size: 20L; Grade: AR (ACS); Concentration: 36.5-38%; CAS: 50-00-0; Hazardous: Y; Container Material: Cubitainer; Formula: HCHO; Formula Weight: 30.03; Suitable for Histological Applications