Learn More
Sigma Aldrich Fine Chemicals Biosciences FORMALDEHYDE SOLUTION 37 WT. %
Catalog No. NC1626119
Encompass
Formaldehyde solution ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Catalog No. NC1626119 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2525494X100ML
Formaldehyde solution ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)