Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Formaldehyde Solution

Catalog No. 503081492 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
1 L
4 L
6 x 500 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1492 6 x 500 mL
25-254-9500ML 500 mL
25-254-925ML 25 mL
25-254-9100ML 100 mL
25-254-91L 1 L
25-254-94L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Danger
Catalog No. 50-308-1492 Supplier Sigma Aldrich Supplier No. 2525496X500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Formaldehyde (formalin) is produced by oxidation of methanol. It is made of 37% formaldehyde and impurities such as methanol, small amounts of formic acid, aldehydes and ketones. It is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA.Formaldehyde solution has been used for cross-linking/fixing of cells in ChIP (chromatin immunoprecipitation) assay. It has been used for fixing of cells for imaging.The article number 252549-4X2.5L will be discontinued. Please order the single bottle 252549-2.5L which is physically identical with the same exact specifications.The article number 252549-6X1L will be discontinued. Please order the single bottle 252549-1L which is physically identical with the same exact specifications.

Specifications

CAS 50-00-0
Density 1.09 g/mL at 25°C
Molecular Formula CH2O
Refractive Index n20/D 1.3765
Linear Formula HCHO
MDL Number MFCD00003274
Quantity 6 x 500 mL
Synonym Formalin
Molecular Weight (g/mol) 30.03 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.