Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Formaldehyde Solution

Catalog No. 1551225LR Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-512-25LR 2.5 L
15-512-1LR 1 L
15-512-5LR 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 15-512-25LR Supplier Sigma Aldrich Supplier No. 155122.5LR
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Formaldehyde solution commercially formaldehyde solution is an aqueous solution containing 37% formaldehyde and 8-10% methanol. Formaldehyde solution can be activated by adding the catalytic amount of lanthanide triflate. This activated formaldehyde solution was employed for the smooth hydroxymethylation reaction of silyl enol ethers. Formaldehyde solution reacts with silyl enol ethers to afford the corresponding hydroxymethylated adducts in high yields.Formaldehyde solution was used to prepare the formaldehyde stock solution for the development of transparent sol-gels with entrapped sensitive and selective reagents for the detection of formaldehyde in air. It may be used in the preparation of nanocrystalline metal (Cu, Hg, Zn, Bi, Pb) sulfides having different shapes and different particle sizes.The article number 15512-4X2.5L-R will be discontinued. Please order the single bottle 15512-2.5L-R which is physically identical with the same exact specifications.The article number 15512-6X1L-R will be discontinued. Please order the single bottle 15512-1L-R which is physically identical with the same exact specifications.

Specifications

CAS 50-00-0
Density 1.09 g/mL at 25°C
Molecular Formula CH2O
Linear Formula HCHO
MDL Number MFCD00003274
Quantity 2.5 L
Synonym Formalin
Molecular Weight (g/mol) 30.03 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.