Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde Solution, ACS, Suitable for Histological Applications, Macron Fine Chemicals™

Catalog No. MK501602 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK501602 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK501602 Supplier Avantor Supplier No. 501602

missing translation for 'validMainframeMsg'

Suitable for histological applications

macron

Specifications

CAS 50-00-0
Packaging Poly Bottle
Quantity 500 mL
Molecular Formula CH2O
MDL Number MFCD00003274
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Molecular Weight (g/mol) 30.03
PubChem CID 712
ChEBI CHEBI:16842
Formula Weight 30.03
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.