Learn More
Supelco Inc Formaldehyde solution 500ml
Catalog No. NC9759988
Encompass
Formaldehyde (formalin) is produced by oxidation of methanol. It is made of 37% formaldehyde and impurities such as methanol, small amounts of formic acid, aldehydes and ketones. It is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. Application:Formaldehyde solution has been used as a fixing agent to fix cells during immunofluorescence imaging and for cross-linking cells during chromatin immunoprecipitation (ChIP) assay. Formaldehyde solution has been used for cross-linking/fixing of cells in ChIP (chromatin immunoprecipitation) assay. It has been used for fixing of cells for imaging. 500ml
Catalog No. NC9759988 Supplier Supelco Inc Supplier No. 252549500ML
Add to Cart
Add to Cart
Formaldehyde (formalin) is produced by oxidation of methanol. It is made of 37% formaldehyde and impurities such as methanol, small amounts of formic acid, aldehydes and ketones. It is used as a denaturant in formaldehyde-agarose gel electrophoresis of RNA. Application:Formaldehyde solution has been used as a fixing agent to fix cells during immunofluorescence imaging and for cross-linking cells during chromatin immunoprecipitation (ChIP) assay. Formaldehyde solution has been used for cross-linking/fixing of cells in ChIP (chromatin immunoprecipitation) assay. It has been used for fixing of cells for imaging. 500ml