Learn More
Sigma Aldrich Fine Chemicals Biosciences formaldehyde solution, 4L
Catalog No. 502898859
Encompass
This product is 37wt% SOLUTIONon of formaldehyde in water. It contains 10-15% methanol as stabilizer.
Catalog No. 50-289-8859 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F16354L
This product is 37wt% SOLUTIONon of formaldehyde in water. It contains 10-15% methanol as stabilizer.