Learn More
Sigma Aldrich Fine Chemicals Biosciences formaldehyde solution cont
Catalog No. 502899628
Encompass
This product is 37wt% SOLUTIONon of formaldehyde in water. It contains 10-15% methanol as stabilizer.
Catalog No. 50-289-9628 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F163525ML
This product is 37wt% SOLUTIONon of formaldehyde in water. It contains 10-15% methanol as stabilizer.