Learn More
Marketlab Formaldehyde Spill Response Hard Case
Catalog No. 22505306
Change view
Click to view available options
Type:
Spill Kit
Catalog No. Type
22-505-306 Spill Kit
1 options
Catalog No. 22-505-306 Supplier Marketlab Supplier No. 15244
Add to Cart
Add to Cart
Features FSC-1 to help reduce formaldehyde gas exposure during spills.
  • 11 oz bottle FSC-1
  • Safety goggles
  • Respirator
  • Impervious gown
  • Latex gloves
  • Pick-up scoops and scrapers
  • Large wiper towel
  • Disposal bags
  • Plastic case
Specifications
Type Spill Kit