Learn More
Electron Microscopy Sciences NeutraTest™ Strips
SDP
Catalog No. 501928594
Encompass_Preferred
These residual low range Formaldehyde test strips are optimized for the NeutraGreen liquid neutralizer. Immerse strip in treated waste solution and results will be rendered in 90 seconds. More efficient and less costly than conventional test kits.
Catalog No. 50-192-8594 Supplier Electron Microscopy Sciences Supplier No. 24147
These residual low range Formaldehyde test strips are optimized for the NeutraGreen liquid neutralizer. Immerse strip in treated waste solution and results will be rendered in 90 seconds. More efficient and less costly than conventional test kits.