Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formaldehyde, Unbuffered, Certified, 10% (v/v), LabChem™

Catalog No. p-5389150
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC146602 1 L
LC146604 4 L
LC146606 10 L
LC146605 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. LC146602 Supplier LabChem, Inc. Supplier No. LC146602
Only null left
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Formaldehyde
Name Note Unbuffered
CAS 7732-18-5, 50-00-0, 67-56-1
CAS Max % 95.05, 3.7, 1.25
Decomposition Information Carbon monoxide; Carbon dioxide
Color Clear, colorless
Packaging Poly Bottle
Linear Formula HCHO
Quantity 1 L
Synonym Formalin
Formula Weight 30.05
Percent Purity 10% (v/v)
Grade Certified
Physical Form Liquid
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.