Learn More
CHEMetrics™ by AquaPhoenix Formaldehyde Water Quality Visual Test Kit
SDP
Catalog No. 13299709
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-709 30
1 options
Catalog No. 13-299-709 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K4605
Add to Cart
Add to Cart
Formaldehyde visual test kit supplied in a plastic case. Contains everything needed to perform 30 tests.
  • K-4605 Kit comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): Refill, Low and High Range Comparators, Activator Solutions, 25 mL sample cup, and instructions
Specifications
Analytes Formaldehyde
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0 to 1 and 1 to 10 ppm
Type Water Quality Visual Test Kit