Learn More
Electron Microscopy Sciences Formaldehyde/Zinc Fixative 4 LT
SDP
Catalog No. 50980479
Encompass_Preferred
Prepared: Ready-to-use. Preserves fine nuclear details and good for immunostaining. Contains no mercury. Can be used with automated tissue processors. Works well with standard and special stains because zinc is a stain enhancing mordant.
Catalog No. 50-980-479 Supplier Electron Microscopy Sciences Supplier No. 1567504
Prepared: Ready-to-use. Preserves fine nuclear details and good for immunostaining. Contains no mercury. Can be used with automated tissue processors. Works well with standard and special stains because zinc is a stain enhancing mordant.